Gene Transcripts

Gene Symbol
Eri1
Entrez Gene ID
361159
Gene Name
exoribonuclease 1
Gene Aliases
RGD1308378, Thex1
Chromosome Location
Chr. 16: 60307148 - 60327237 on Build Rnor_6.0
UniGene ID
Rn.17469
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014143.1 NP_001014165.1 7 1201
GenBank mRNA BC089828.1 - 7 1201

View Full Product Details