Gene Transcripts

Gene Symbol
Klk1b3
Entrez Gene ID
18050
Gene Name
kallikrein 1-related peptidase b3
Gene Aliases
Ngfg, mGk-3
Chromosome Location
Chr. 7: 44198191 - 44202351 on Build GRCm38
UniGene ID
Mm.439740
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008693.2 NP_032719.1 3 294
GenBank mRNA BC132657.1 - 3 294
BC132659.1 - 3 294 40130936
X01389.1 - 3 255

View Full Product Details