Gene Transcripts
- Gene Symbol
- H2-M10.3
- Entrez Gene ID
- 110696
- Gene Name
- histocompatibility 2, M region locus 10.3
- Gene Aliases
- 5.3H
- Chromosome Location
- Chr. 17: 36365004 - 36368417 on Build GRCm38
- UniGene ID
- Mm.303091
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY211080.1 |
- |
6 |
1149 |
|
View Full Product Details