Gene Transcripts

Gene Symbol
Gpr34
Entrez Gene ID
554353
Gene Name
G protein-coupled receptor 34
Gene Aliases
Chromosome Location
Chr. X: 10022986 - 10043500 on Build Rnor_6.0
UniGene ID
Rn.163444
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256675.2 XP_006256737.1 4 3673
XM_006256676.2 XP_006256738.1 4 1590

View Full Product Details