Gene Transcripts

Gene Symbol
NMS
Entrez Gene ID
129521
Gene Name
neuromedin S
Gene Aliases
Chromosome Location
Chr. 2: 100470482 - 100483280 on Build GRCh38
UniGene ID
Hs.567676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011717.1 NP_001011717.1 3 178
GenBank mRNA AB164464.1 - 3 178

View Full Product Details