Gene Transcripts

Gene Symbol
C14orf28
Entrez Gene ID
122525
Gene Name
chromosome 14 open reading frame 28
Gene Aliases
DRIP-1, DRIP1, c14_5270
Chromosome Location
Chr. 14: 44897304 - 44907257 on Build GRCh38
UniGene ID
Hs.82098
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017923.1 NP_001017923.1 5 2525
XM_011536408.1 XP_011534710.1 4 1826
GenBank mRNA AK126985.1 - 3 3118

View Full Product Details