Gene Transcripts

Gene Symbol
Hgfac
Entrez Gene ID
58947
Gene Name
HGF activator
Gene Aliases
Chromosome Location
Chr. 14: 80966902 - 80973559 on Build Rnor_6.0
UniGene ID
Rn.144587
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053320.1 NP_445772.1 11 1470
XM_008770352.1 XP_008768574.1 11 1573
GenBank mRNA BC089871.1 - 11 1470

View Full Product Details