Gene Transcripts

Gene Symbol
Vom1r42
Entrez Gene ID
494303
Gene Name
vomeronasal 1 receptor 42
Gene Aliases
V1rg16
Chromosome Location
Chr. 1: 67301831 - 67302751 on Build Rnor_6.0
UniGene ID
Rn.215803
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008959.1 NP_001008959.1 1 775
GenBank mRNA AY510306.1 - 1 775

View Full Product Details