Gene Transcripts

Gene Symbol
EBLN2
Entrez Gene ID
55096
Gene Name
endogenous Bornavirus-like nucleoprotein 2
Gene Aliases
EBLN-2
Chromosome Location
Chr. 3: 73061659 - 73063337 on Build GRCh38
UniGene ID
Hs.658858
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018029.4 NP_060499.3 1 1364
GenBank mRNA AK001075.1 - 1 751
AY036895.1 - 1 942
BC051316.1 - 1 1364 6214587
BC064501.1 - 1 1364 6215884

View Full Product Details