Gene Transcripts
- Gene Symbol
- Vps52
- Entrez Gene ID
- 224705
- Gene Name
- vacuolar protein sorting 52 (yeast)
- Gene Aliases
- ARE1, D130068D18, D430041K17Rik, SAC2, Sacm2l, tclw5
- Chromosome Location
- Chr. 17: 33955785 - 33966495 on Build GRCm38
- UniGene ID
- Mm.448614
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042520.1 |
- |
20 |
2969 |
|
AK051732.1 |
- |
20 |
3085 |
|
View Full Product Details