Gene Transcripts

Gene Symbol
NMS
Entrez Gene ID
497196
Gene Name
neuromedin S
Gene Aliases
Chromosome Location
Chr. 9: 45605552 - 45616912 on Build Rnor_6.0
UniGene ID
Rn.137581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012233.1 NP_001012233.1 485
GenBank mRNA AB164465.1 - 485

View Full Product Details