Gene Transcripts

Gene Symbol
Ahrr
Entrez Gene ID
498999
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
Chromosome Location
Chr. 1: 31603297 - 31698329 on Build Rnor_6.0
UniGene ID
Rn.145524
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024285.1 NP_001019456.1 11 1823
GenBank mRNA AB174900.2 - 11 1823
AY367561.1 - 11 1823

View Full Product Details