Gene Transcripts

Gene Symbol
TMEM57
Entrez Gene ID
55219
Gene Name
transmembrane protein 57
Gene Aliases
MACOILIN
Chromosome Location
Chr. 1: 25430858 - 25500209 on Build GRCh38
UniGene ID
Hs.189782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282564.1 NP_001269493.1 1, 2 315
NM_018202.5 NP_060672.2 1, 2 315
XM_005245931.2 XP_005245988.1 1, 2 276
XM_011541704.2 XP_011540006.1 1, 2 269
GenBank mRNA AK001609.1 - 1, 2 226
AK311472.1 - 1, 2 276
AY845015.1 - 1, 2 245
AY845035.1 - 1, 2 183
BC032427.1 - 1, 2 183 5246967
DB455940.1 - 1, 2 315

View Full Product Details