Gene Transcripts

Gene Symbol
MSL3P1
Entrez Gene ID
151507
Gene Name
male-specific lethal 3 homolog (Drosophila) pseudogene 1
Gene Aliases
MSL3L2
Chromosome Location
Chr. 2: 233865437 - 233868409 on Build GRCh38
UniGene ID
Hs.355809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024322.1 1 74
GenBank mRNA AK301707.1 - 1 58
AK316372.1 - 1 25
BI831020.1 - 1 44
DB457661.1 - 1 74
DC394932.1 - 1 25

View Full Product Details