Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018229.3 | NP_060699.3 | 2 | 930 | |
XM_006720196.2 | XP_006720259.1 | 2 | 559 | ||
XM_011536940.2 | XP_011535242.1 | 3 | 902 | ||
GenBank mRNA | AK001675.1 | - | 2 | 775 | |
AK225345.1 | - | 2 | 777 | ||
AK303999.1 | - | 3 | 804 | ||
AL136685.1 | - | 2 | 909 | ||
BC013174.1 | - | 2 | 894 | 3924075 |