Gene Transcripts

Gene Symbol
SSPO
Entrez Gene ID
23145
Gene Name
SCO-spondin
Gene Aliases
Chromosome Location
Chr. 7: 149776042 - 149833965 on Build GRCh38
UniGene ID
Hs.632022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198455.2 NP_940857.2 101 15221
GenBank mRNA AK093431.1 - 10 2170
AK123170.1 - 17 2236
AK124542.1 - 8 3106

View Full Product Details