Gene Transcripts

Gene Symbol
CWF19L1
Entrez Gene ID
55280
Gene Name
CWF19-like 1, cell cycle control (S. pombe)
Gene Aliases
C19L1, SCAR17, hDrn1
Chromosome Location
Chr. 10: 100232296 - 100267681 on Build GRCh38
UniGene ID
Hs.215502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303404.1 NP_001290333.1 3 258
NM_001303405.1 NP_001290334.1 3 303
NM_001303407.1 NP_001290336.1 3 242
NM_018294.5 NP_060764.3 3 258
GenBank mRNA AK001860.1 - 3 204
AK023984.1 - 3 177
AK097895.1 - 3 241
AK225590.1 - 3 216
AL832515.1 - 3 215
BC008746.2 - 3 199 3345869
DC299678.1 - 3 258
KU178692.1 - 3 170
KU178693.1 - 3 154

View Full Product Details