Gene Transcripts
- Gene Symbol
- FBXO6
- Entrez Gene ID
- 26270
- Gene Name
- F-box protein 6
- Gene Aliases
- FBG2, FBS2, FBX6, Fbx6b
- Chromosome Location
- Chr. 1: 11664093 - 11674354 on Build GRCh38
- UniGene ID
- Hs.464419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018438.5 |
NP_060908.1
|
4, 5 |
678 |
|
XM_005263448.4 |
XP_005263505.1
|
4, 5 |
618 |
|
XM_005263449.4 |
XP_005263506.1
|
4, 5 |
565 |
|
XM_005263451.4 |
XP_005263508.1
|
4, 5 |
618 |
|
XM_006710571.3 |
XP_006710634.1
|
4, 5 |
646 |
|
XM_011541232.2 |
XP_011539534.1
|
4, 5 |
612 |
|
XM_017001007.1 |
XP_016856496.1
|
5, 6 |
680 |
|
GenBank mRNA |
AF129536.1 |
- |
4, 5 |
647 |
|
AF233223.1 |
- |
3, 4 |
523 |
|
AK314989.1 |
- |
4, 5 |
678 |
|
BC020880.1 |
- |
4, 5 |
620 |
4614957 |
View Full Product Details