Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001002273.2 | NP_001002273.1 | 7 | 947 | |
NM_001002274.2 | NP_001002274.1 | 7 | 950 | ||
NM_001002275.2 | NP_001002275.1 | 8 | 1004 | ||
NM_001190828.1 | NP_001177757.1 | 7 | 986 | ||
NM_004001.4 | NP_003992.3 | 8 | 1007 | ||
XM_017000671.1 | XP_016856160.1 | 9 | 971 | ||
XM_017000672.1 | XP_016856161.1 | 8 | 914 | ||
GenBank mRNA | AB209585.1 | - | 6 | 4128 | |
AF543826.1 | - | 8 | 960 | ||
AK308150.1 | - | 7 | 877 | ||
BC031992.1 | - | 8 | 917 | 4752609 | |
BC141826.1 | - | 7 | 860 | 40148276 | |
BC146678.1 | - | 7 | 865 | 40148906 | |
BC148273.1 | - | 8 | 902 | 8860219 | |
CR407635.1 | - | 7 | 823 | ||
M28696.1 | - | 7 | 864 | ||
M31933.1 | - | 7 | 939 | ||
M31934.1 | - | 7 | 862 | ||
M31935.1 | - | 8 | 830 | ||
U87560.1 | - | 8 | 880 | ||
U87561.1 | - | 8 | 880 | ||
U87562.1 | - | 8 | 880 | ||
U87563.1 | - | 8 | 880 | ||
U87564.1 | - | 7 | 823 | ||
U87565.1 | - | 7 | 823 | ||
U87566.1 | - | 7 | 823 | ||
U87567.1 | - | 7 | 823 | ||
U87568.1 | - | 7 | 823 | ||
U87569.1 | - | 7 | 823 | ||
U87570.1 | - | 8 | 877 | ||
U87571.1 | - | 7 | 820 | ||
U87572.1 | - | 8 | 877 | ||
U87573.1 | - | 8 | 880 | ||
U87574.1 | - | 7 | 823 | ||
U87575.1 | - | 7 | 823 | ||
U87576.1 | - | 8 | 880 | ||
U87577.1 | - | 8 | 877 | ||
X17653.1 | - | 7 | 892 | ||
X52473.1 | - | 7 | 868 |