Gene Transcripts

Gene Symbol
FAM21A
Entrez Gene ID
387680
Gene Name
family with sequence similarity 21 member A
Gene Aliases
FAM21B, bA56A21.1, bA98I6.1
Chromosome Location
Chr. 10: 50067888 - 50133509 on Build GRCh38
UniGene ID
Hs.365286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005751.2 NP_001005751.1 27, 28 2987
XM_005269806.3 XP_005269863.2 26, 27 2863
XM_005269807.3 XP_005269864.2 26, 27 2848
XM_005269810.3 XP_005269867.2 9, 10 1080
XM_006717831.3 XP_006717894.2 26, 27 2782
XM_017016227.1 XP_016871716.1 26, 27 2812
XM_017016230.1 XP_016871719.1 26, 27 2800
XM_017016233.1 XP_016871722.1 24, 25 2510
XM_017016234.1 XP_016871723.1 26, 27 2870
XM_017016235.1 XP_016871724.1 18, 19 1886
GenBank mRNA AK001686.1 - 24, 25 2639
AK304116.1 - 27, 28 2923
BC082258.1 - 27, 28 2914 6278433
CN314884.1 - 4, 5 312

Gene Symbol
FAM21C
Entrez Gene ID
253725
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, VPEF
Chromosome Location
Chr. 10: 45727170 - 45792964 on Build GRCh38
UniGene ID
Hs.365286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011539566.1 XP_011537868.1 27, 28 2954
XM_011539567.1 XP_011537869.1 26, 27 2882
XM_011539568.1 XP_011537870.1 26, 27 2867
XM_011539569.1 XP_011537871.1 26, 27 2801
XM_011539570.2 XP_011537872.1 26, 27 2843
XM_011539573.2 XP_011537875.1 9, 10 1080
XM_017016014.1 XP_016871503.1 26, 27 2831
XM_017016017.1 XP_016871506.1 25, 26 2795
XM_017016019.1 XP_016871508.1 25, 26 2729
XM_017016021.1 XP_016871510.1 27, 28 2882
XM_017016024.1 XP_016871513.1 15, 16 1696
XM_017016025.1 XP_016871514.1 16, 17 1767
GenBank mRNA AK001545.1 - 9, 10 1148
AK293929.1 - 22, 23 2438

View Full Product Details