Gene Transcripts

Gene Symbol
C2orf76
Entrez Gene ID
130355
Gene Name
chromosome 2 open reading frame 76
Gene Aliases
AIM29
Chromosome Location
Chr. 2: 119281472 - 119366879 on Build GRCh38
UniGene ID
Hs.99488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017927.3 NP_001017927.2 7 1091
NM_001322329.1 NP_001309258.1 7 1012
NM_001322330.1 NP_001309259.1 6 888
NM_001322331.1 NP_001309260.1 6 541
NM_001322332.1 NP_001309261.1 5 490
GenBank mRNA BC047365.1 - 7 1012
BC062434.1 - 5 523
BC105805.1 - 6 545 6496685

View Full Product Details