Gene Transcripts

Gene Symbol
EME2
Entrez Gene ID
197342
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
SLX2B, gs125
Chromosome Location
Chr. 16: 1773222 - 1776714 on Build GRCh38
UniGene ID
Hs.7247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001257370.1 NP_001244299.1 3 417
XM_017023025.1 XP_016878514.1 3 339
GenBank mRNA AK074080.2 - 3 319
EF452422.1 - 3 417

View Full Product Details