Gene Transcripts

Gene Symbol
SPANXN1
Entrez Gene ID
494118
Gene Name
SPANX family member N1
Gene Aliases
CT11.6
Chromosome Location
Chr. X: 145247587 - 145256208 on Build GRCh38
UniGene ID
Hs.551270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009614.2 NP_001009614.1 2 392
GenBank mRNA DQ336118.1 - 2 392
DQ336119.1 - 2 392
DQ336120.1 - 2 392
DQ336121.1 - 2 392

View Full Product Details