Gene Transcripts

Gene Symbol
CTRB2
Entrez Gene ID
440387
Gene Name
chymotrypsinogen B2
Gene Aliases
Chromosome Location
Chr. 16: 75204096 - 75207174 on Build GRCh38
UniGene ID
Hs.610926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025200.3 NP_001020371.3 7 865
GenBank mRNA AK131056.1 - 852
AK225933.1 - 7 852
AK291822.1 - 7 850
BC073145.1 - 841 5747536

View Full Product Details