Gene Transcripts

Gene Symbol
Pcdhgc3
Entrez Gene ID
116782
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
Chromosome Location
Chr. 18: 30954552 - 30971113 on Build Rnor_6.0
UniGene ID
Rn.8628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053943.1 NP_446395.1 1 1418
GenBank mRNA AY574028.1 - 1 1418

View Full Product Details