Gene Transcripts

Gene Symbol
Cfap20
Entrez Gene ID
307642
Gene Name
cilia and flagella associated protein 20
Gene Aliases
Gtl3
Chromosome Location
Chr. 19: 10024938 - 10038838 on Build Rnor_6.0
UniGene ID
Rn.6708
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037978.1 NP_001033067.1 4 568
XM_006255083.1 XP_006255145.1 4 654
GenBank mRNA BC099768.1 - 4 568

View Full Product Details