Gene Transcripts

Gene Symbol
Ppef1
Entrez Gene ID
317498
Gene Name
protein phosphatase, EF-hand calcium binding domain 1
Gene Aliases
Chromosome Location
Chr. X: 35822687 - 35947690 on Build Rnor_6.0
UniGene ID
Rn.103108
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034935.1 NP_001030107.1 9 1086
GenBank mRNA BC104696.1 - 9 1086

View Full Product Details