Gene Transcripts

Gene Symbol
Clul1
Entrez Gene ID
367345
Gene Name
clusterin-like 1 (retinal)
Gene Aliases
Chromosome Location
Chr. 9: 121946316 - 121972055 on Build Rnor_6.0
UniGene ID
Rn.136023
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033071.1 NP_001028243.1 2 117
GenBank mRNA AY655707.1 - 2 117

View Full Product Details