Gene Transcripts
- Gene Symbol
- ANKRD2
- Entrez Gene ID
- 26287
- Gene Name
- ankyrin repeat domain 2
- Gene Aliases
- ARPP
- Chromosome Location
- Chr. 10: 97572441 - 97583884 on Build GRCh38
- UniGene ID
- Hs.73708
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001129981.2 |
NP_001123453.1
|
1, 2 |
428 |
|
NM_001291218.1 |
NP_001278147.1
|
1, 2 |
428 |
|
NM_001291219.1 |
NP_001278148.1
|
1, 2 |
428 |
|
NM_020349.3 |
NP_065082.2
|
1, 2 |
428 |
|
GenBank mRNA |
AB058599.1 |
- |
1, 2 |
90 |
|
AJ249975.1 |
- |
|
18 |
|
AJ304805.1 |
- |
1, 2 |
90 |
|
AJ583444.1 |
- |
1, 2 |
428 |
|
AK056990.1 |
- |
1, 2 |
370 |
|
AK314763.1 |
- |
1, 2 |
90 |
|
BC020817.1 |
- |
1, 2 |
90 |
|
BC107759.1 |
- |
1, 2 |
331 |
4096546 |
DA893829.1 |
- |
1, 2 |
370 |
|
View Full Product Details