Gene Transcripts

Gene Symbol
Wdr91
Entrez Gene ID
101240
Gene Name
WD repeat domain 91
Gene Aliases
9530020G05Rik, AI987683, AU018665
Chromosome Location
Chr. 6: 34879062 - 34910881 on Build GRCm38
UniGene ID
Mm.152120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013366.1 NP_001013384.1 15 2499
GenBank mRNA AK035345.1 - 1 673
BC043682.1 - 10 1768
BC055046.1 - 15 2499 6488548

View Full Product Details