Gene Transcripts
- Gene Symbol
- Dhrsx
- Entrez Gene ID
- 236082
- Gene Name
- dehydrogenase/reductase (SDR family) X chromosome
- Gene Aliases
- C80877, D4Ertd296e, Dhrsxy, Pscad
- Chromosome Location
- Chr. 4|NT_166291: 15800 - 36213 on Build GRCm38
- UniGene ID
- Mm.305345
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK157461.1 |
- |
7 |
1721 |
|
View Full Product Details