Gene Transcripts
- Gene Symbol
- Cyp4a31
- Entrez Gene ID
- 666168
- Gene Name
- cytochrome P450, family 4, subfamily a, polypeptide 31
- Gene Aliases
- Chromosome Location
- Chr. 4: 115563626 - 115579015 on Build GRCm38
- UniGene ID
- Mm.482086
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC013476.1 |
- |
11 |
1819 |
|
View Full Product Details