Gene Transcripts

Gene Symbol
LCE3E
Entrez Gene ID
353145
Gene Name
late cornified envelope 3E
Gene Aliases
LEP17
Chromosome Location
Chr. 1: 152565654 - 152566770 on Build GRCh38
UniGene ID
Hs.490208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178435.3 NP_848522.1 2 501
XM_011509484.1 XP_011507786.1 2 597

View Full Product Details