Gene Transcripts
- Gene Symbol
- LOC100048884
- Entrez Gene ID
- 100048884
- Gene Name
- novel member of the major urinary protein (Mup) gene family
- Gene Aliases
- Mup6
- Chromosome Location
- Chr. 4: 61670177 - 61674098 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC055353.1 |
- |
|
667 |
|
- Gene Symbol
- Mup1
- Entrez Gene ID
- 17840
- Gene Name
- major urinary protein 1
- Gene Aliases
- Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
- Chromosome Location
- Chr. 4: 60498012 - 60501960 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
- Gene Symbol
- Mup10
- Entrez Gene ID
- 100039008
- Gene Name
- major urinary protein 10
- Gene Aliases
- 2610016E04Rik, Mup8
- Chromosome Location
- Chr. 4: 60578260 - 60582155 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK011413.1 |
- |
4 |
739 |
|
- Gene Symbol
- Mup2
- Entrez Gene ID
- 17841
- Gene Name
- major urinary protein 2
- Gene Aliases
- AA589603, Mup-2, Mup4
- Chromosome Location
- Chr. 4: 60135913 - 60139872 on Build GRCm38
- UniGene ID
- Mm.335875
- Species
- Mus musculus
- Gene Symbol
- Mup3
- Entrez Gene ID
- 17842
- Gene Name
- major urinary protein 3
- Gene Aliases
- MUP15, MUPIII, Mup-3, Mup25
- Chromosome Location
- Chr. 4: 62083476 - 62087370 on Build GRCm38
- UniGene ID
- Mm.250267
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M27608.1 |
- |
6 |
662 |
|
View Full Product Details