Gene Transcripts

Gene Symbol
Fgfbp3
Entrez Gene ID
72514
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
2610306H15Rik, Fgf-bp3
Chromosome Location
Chr. 19: 36917550 - 36919599 on Build GRCm38
UniGene ID
Mm.41674
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028263.1 NP_082539.2 2 1263
GenBank mRNA AK011996.1 - 1 1073
AK032142.1 - 1 361
AK167949.1 - 2 1281
BC055778.1 - 2 1263 6403098

View Full Product Details