Gene Transcripts

Gene Symbol
Ip6k2
Entrez Gene ID
76500
Gene Name
inositol hexaphosphate kinase 2
Gene Aliases
1500005N04Rik, AW050208, Ihpk2
Chromosome Location
Chr. 9: 108783764 - 108806333 on Build GRCm38
UniGene ID
Mm.276336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029634.2 NP_083910.2 6 1574
GenBank mRNA AK050593.1 - 6 1432
BC039922.1 - 6 1585 2609803
BC096640.1 - 460

View Full Product Details