Gene Transcripts

Gene Symbol
Vmn2r7
Entrez Gene ID
319217
Gene Name
vomeronasal 2, receptor 7
Gene Aliases
4933425M15Rik
Chromosome Location
Chr. 3: 64690660 - 64719602 on Build GRCm38
UniGene ID
Mm.246646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175674.3 NP_783605.2 1 190
GenBank mRNA AK030224.1 - 1289

View Full Product Details