Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001711.2 | NP_001001711.1 | 1 | 976 | |
GenBank mRNA | AF429971.1 | - | 1 | 972 | |
AK093336.1 | - | 1 | 956 | ||
BC021172.2 | - | 1 | 917 | 4822028 | |
BC021710.1 | - | 1 | 920 | 4826837 | |
BC022017.2 | - | 1 | 943 | 4825124 | |
BC022018.1 | - | 1 | 964 | 4825958 |