Gene Transcripts

Gene Symbol
DIABLO
Entrez Gene ID
56616
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
DFNA64, SMAC
Chromosome Location
Chr. 12: 122207662 - 122227534 on Build GRCh38
UniGene ID
Hs.169611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278302.1 NP_001265231.1 5 1392
NM_001278303.1 NP_001265232.1 5 1420
NM_001278304.1 NP_001265233.1 5 2022
NM_001278342.1 NP_001265271.1 5 1421
NM_019887.5 NP_063940.1 7 1817
NM_138930.3 NP_620308.1 6 1524
GenBank mRNA AF262240.1 - 6 1018
AK001399.1 - 5 2026
AK024768.1 - 6 1026
AK057778.1 - 5 894
AK222664.1 - 6 1015
BC004417.1 - 4 832
BC011909.2 - 6 1022 4137792
BC046209.1 - 7 1739
BC095468.1 - 6 1008 30557745

View Full Product Details