Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001302378.1 | NP_001289307.1 | 4 | 695 | |
NM_022340.3 | NP_071735.2 | 5 | 851 | ||
XM_005265384.4 | XP_005265441.1 | 5 | 814 | ||
XM_005265385.4 | XP_005265442.1 | 4 | 669 | ||
XM_017007023.1 | XP_016862512.1 | 4 | 687 | ||
XM_017007024.1 | XP_016862513.1 | 6 | 997 | ||
XM_017007025.1 | XP_016862514.1 | 5 | 856 | ||
XM_017007026.1 | XP_016862515.1 | 4 | 701 | ||
XM_017007027.1 | XP_016862516.1 | 5 | 842 | ||
XM_017007028.1 | XP_016862517.1 | 5 | 842 | ||
GenBank mRNA | AB209673.1 | - | 5 | 811 | |
AK291288.1 | - | 4 | 686 | ||
AK301735.1 | - | 4 | 687 | ||
AL833545.1 | - | 5 | 844 | ||
AY009133.1 | - | 2 | 228 | ||
BC106940.2 | - | 2 | 228 | 40034008 |