Gene Transcripts
- Gene Symbol
- RNASEH2B
- Entrez Gene ID
- 79621
- Gene Name
- ribonuclease H2 subunit B
- Gene Aliases
- AGS2, DLEU8
- Chromosome Location
- Chr. 13: 50909678 - 50970462 on Build GRCh38
- UniGene ID
- Hs.306291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142279.2 |
NP_001135751.1
|
2, 3 |
543 |
|
NM_024570.3 |
NP_078846.2
|
2, 3 |
543 |
|
XM_006719867.3 |
XP_006719930.1
|
2, 3 |
232 |
|
XM_011535229.1 |
XP_011533531.1
|
2, 3 |
465 |
|
XM_011535230.2 |
XP_011533532.1
|
2, 3 |
465 |
|
XM_011535231.2 |
XP_011533533.1
|
2, 3 |
465 |
|
XM_011535233.2 |
XP_011533535.1
|
2, 3 |
213 |
|
XM_017020747.1 |
XP_016876236.1
|
2, 3 |
465 |
|
GenBank mRNA |
AK021774.1 |
- |
2, 3 |
430 |
|
AK223340.1 |
- |
2, 3 |
430 |
|
AY764036.1 |
- |
2, 3 |
451 |
|
BC001397.2 |
- |
2, 3 |
253 |
|
BC005088.1 |
- |
2, 3 |
253 |
|
BC007332.2 |
- |
2, 3 |
253 |
|
BC010174.2 |
- |
2, 3 |
253 |
|
BC036744.1 |
- |
2, 3 |
414 |
5195426 |
BX382357.2 |
- |
2, 3 |
421 |
|
DB221997.1 |
- |
2, 3 |
465 |
|
DT216141.1 |
- |
2, 3 |
465 |
|
View Full Product Details