Gene Transcripts
- Gene Symbol
- NKAIN1
- Entrez Gene ID
- 79570
- Gene Name
- Na+/K+ transporting ATPase interacting 1
- Gene Aliases
- FAM77C
- Chromosome Location
- Chr. 1: 31179270 - 31239887 on Build GRCh38
- UniGene ID
- Hs.470259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024522.2 |
NP_078798.2
|
5, 6 |
870 |
|
XM_017002320.1 |
XP_016857809.1
|
4, 5 |
464 |
|
GenBank mRNA |
AK022712.1 |
- |
4, 5 |
442 |
|
AK298874.1 |
- |
3, 4 |
361 |
|
AK299507.1 |
- |
5, 6 |
542 |
|
AL832547.1 |
- |
2, 3 |
135 |
|
BC096707.1 |
- |
4, 5 |
428 |
40022279 |
BC098173.1 |
- |
4, 5 |
428 |
40022282 |
BC098249.1 |
- |
4, 5 |
428 |
40022276 |
BC131518.1 |
- |
4, 5 |
428 |
40022275 |
View Full Product Details