Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133264.4 | NP_573571.1 | 4 | 453 | |
XM_005257083.2 | XP_005257140.1 | 4 | 455 | ||
XM_005257084.2 | XP_005257141.1 | 4 | 413 | ||
XM_011524413.1 | XP_011522715.1 | 3 | 335 | ||
GenBank mRNA | AB043786.1 | - | 3 | 213 | |
AF370396.1 | - | 4 | 377 | ||
AJ431177.1 | - | 3 | 213 | ||
AK026913.1 | - | 2 | 45 | ||
AK289578.1 | - | 4 | 413 | ||
AK289779.1 | - | 4 | 472 | ||
AK290301.1 | - | 4 | 449 | ||
AK315836.1 | - | 4 | 433 | ||
AL834239.1 | - | 3 | 322 | ||
BC025965.1 | - | 4 | 409 | ||
BC065551.1 | - | 4 | 390 | 6044222 | |
CN404607.1 | - | 4 | 453 |