Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098502.1 | NP_001091972.1 | 3 | 488 | |
NM_144636.2 | NP_653237.1 | 4 | 658 | ||
GenBank mRNA | AK056271.1 | - | 4 | 609 | |
AK290764.1 | - | 3 | 416 | ||
BC017082.1 | - | 3 | 356 | 3848900 | |
BC033775.1 | - | 3 | 384 | 4538841 |