Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006572.5 | NP_006563.2 | 1, 2 | 500 | |
XM_011524202.1 | XP_011522504.1 | 1, 2 | 294 | ||
GenBank mRNA | AF493902.1 | - | 1, 2 | 291 | |
AK301699.1 | - | 2, 3 | 425 | ||
AK313672.1 | - | 1, 2 | 294 | ||
BC036756.1 | - | 1, 2 | 345 | 5548632 | |
DC335364.1 | - | 1, 2 | 500 | ||
L22075.1 | - | 1, 2 | 332 |