Gene Transcripts
- Gene Symbol
- LRRC75A-AS1
- Entrez Gene ID
- 125144
- Gene Name
- LRRC75A antisense RNA 1
- Gene Aliases
- C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
- Chromosome Location
- Chr. 17: 16438987 - 16470648 on Build GRCh38
- UniGene ID
- Hs.368934
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027159.1 |
|
2, 3 |
505 |
|
NR_027160.1 |
|
3, 4 |
628 |
|
NR_027162.1 |
|
3, 4 |
681 |
|
NR_027163.1 |
|
4, 5 |
415 |
|
NR_027164.1 |
|
4, 5 |
362 |
|
NR_027166.1 |
|
3, 4 |
239 |
|
NR_027168.2 |
|
4, 5 |
331 |
|
NR_027170.1 |
|
4, 5 |
283 |
|
NR_027171.1 |
|
2, 3 |
505 |
|
NR_027172.2 |
|
3, 4 |
208 |
|
NR_027173.1 |
|
3, 4 |
239 |
|
NR_027174.1 |
|
3, 4 |
239 |
|
NR_027175.1 |
|
3, 4 |
239 |
|
NR_027177.1 |
|
3, 4 |
239 |
|
NR_027178.1 |
|
3, 4 |
239 |
|
NR_027179.1 |
|
3, 4 |
239 |
|
NR_027667.1 |
|
3, 4 |
549 |
|
NR_045021.1 |
|
3, 4 |
218 |
|
NR_045022.1 |
|
4, 5 |
357 |
|
NR_045024.1 |
|
2, 3 |
505 |
|
NR_045025.1 |
|
2, 3 |
151 |
|
NR_045026.1 |
|
4, 5 |
415 |
|
NR_045029.1 |
|
3, 4 |
313 |
|
GenBank mRNA |
AK098643.1 |
- |
3, 4 |
189 |
|
AK307982.1 |
- |
3, 4 |
631 |
|
AK311247.1 |
- |
3, 4 |
795 |
|
BC019833.1 |
- |
2, 3 |
165 |
|
BC027986.1 |
- |
3, 4 |
499 |
4940349 |
BC040159.1 |
- |
3, 4 |
478 |
5539041 |
BQ070126.1 |
- |
2, 3 |
243 |
|
BU506593.1 |
- |
|
346 |
|
BU599615.1 |
- |
4, 5 |
369 |
|
BX538291.1 |
- |
4, 5 |
234 |
|
CB150672.1 |
- |
4, 5 |
312 |
|
CD050014.1 |
- |
3, 4 |
212 |
|
CD389053.1 |
- |
3, 4 |
189 |
|
CF126952.1 |
- |
2, 3 |
421 |
|
CR989654.1 |
- |
3, 4 |
292 |
|
DA131201.1 |
- |
3, 4 |
189 |
|
DA193585.1 |
- |
3, 4 |
374 |
|
DA383448.1 |
- |
3, 4 |
189 |
|
DA440445.1 |
- |
4, 5 |
308 |
|
DA485648.1 |
- |
3, 4 |
190 |
|
DA494627.1 |
- |
2, 3 |
456 |
|
DA793334.1 |
- |
2, 3 |
105 |
|
DA802938.1 |
- |
3, 4 |
189 |
|
DA857697.1 |
- |
3, 4 |
189 |
|
DA867958.1 |
- |
4, 5 |
367 |
|
DB258221.1 |
- |
2, 3 |
456 |
|
DW409012.1 |
- |
3, 4 |
206 |
|
View Full Product Details