Gene Transcripts

Gene Symbol
SLC39A12
Entrez Gene ID
221074
Gene Name
solute carrier family 39 member 12
Gene Aliases
LZT-Hs8, ZIP-12, bA570F3.1
Chromosome Location
Chr. 10: 17951839 - 18043292 on Build GRCh38
UniGene ID
Hs.350895
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145195.1 NP_001138667.1 3 579
NM_001282733.1 NP_001269662.1 3 579
NM_001282734.1 NP_001269663.1 2 232
NM_152725.3 NP_689938.2 3 579
GenBank mRNA AK055061.1 - 3 527
AK295294.1 - 2 108
AK303735.1 - 3 264
BC035118.1 - 3 511
BC047635.1 - 3 494 4820920
BC065917.1 - 2 323
BC094700.1 - 3 455 30407123
BC117323.1 - 2 329 40125874
BC143551.1 - 2 328 9052062

View Full Product Details