Gene Transcripts

Gene Symbol
FAM71C
Entrez Gene ID
196472
Gene Name
family with sequence similarity 71 member C
Gene Aliases
Chromosome Location
Chr. 12: 99647750 - 99650114 on Build GRCh38
UniGene ID
Hs.591011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153364.3 NP_699195.1 1 555
GenBank mRNA AK312767.1 - 1 551
BC031221.1 - 1 552 5272209

View Full Product Details