Gene Transcripts

Gene Symbol
SYCP3
Entrez Gene ID
50511
Gene Name
synaptonemal complex protein 3
Gene Aliases
COR1, RPRGL4, SCP3, SPGF4
Chromosome Location
Chr. 12: 101728648 - 101739722 on Build GRCh38
UniGene ID
Hs.506504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177948.1 NP_001171419.1 3 313
NM_001177949.1 NP_001171420.1 3 328
NM_153694.4 NP_710161.1 3 324
XM_005268922.4 XP_005268979.3 3 574
XM_005268924.1 XP_005268981.1 3 312
XM_005268925.1 XP_005268982.1 3 317
XM_005268926.3 XP_005268983.1 3 296
XM_005268927.2 XP_005268984.1 3 285
XM_011538421.2 XP_011536723.2 3 496
XM_017019368.1 XP_016874857.1 3 571
GenBank mRNA AF492003.1 - 3 269
AF517774.1 - 3 267
AK125930.1 - 2 819
BC062662.1 - 3 319 6616222
BQ428625.1 - 3 329

View Full Product Details