Gene Transcripts

Gene Symbol
SLC38A9
Entrez Gene ID
153129
Gene Name
solute carrier family 38 member 9
Gene Aliases
URLC11
Chromosome Location
Chr. 5: 55625845 - 55712344 on Build GRCh38
UniGene ID
Hs.742101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258286.1 NP_001245215.1 3 696
NM_001258287.1 NP_001245216.1 4 800
NM_001282429.1 NP_001269358.1 4 413
NM_173514.3 NP_775785.2 5 546
NR_047649.1 3 696
XM_006714537.2 XP_006714600.1 7 780
XM_006714538.3 XP_006714601.1 6 667
XM_006714539.3 XP_006714602.1 6 710
XM_011543173.1 XP_011541475.1 7 768
XM_011543174.1 XP_011541476.1 8 866
XM_011543176.1 XP_011541478.1 7 780
XM_011543177.1 XP_011541479.1 7 780
XM_011543178.1 XP_011541480.1 7 780
XM_011543180.1 XP_011541482.1 7 780
XM_011543182.1 XP_011541484.1 7 780
XM_011543183.1 XP_011541485.1 7 780
XM_011543184.1 XP_011541486.1 7 780
XM_017009077.1 XP_016864566.1 5 478
XM_017009078.1 XP_016864567.1 5 555
XM_017009079.1 XP_016864568.1 5 478
XM_017009080.1 XP_016864569.1 6 667
XM_017009081.1 XP_016864570.1 5 478
XM_017009082.1 XP_016864571.1 5 478
GenBank mRNA AB105188.1 - 5 478
AK075190.1 - 5 478
AK122957.1 - 4 393
AK127988.1 - 4 440
AK301850.1 - 3 358
BC066891.1 - 5 535 4373965
BC101362.2 - 5 477 40025719
DC371481.1 - 5 546

View Full Product Details